accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2250-55-7

FFFHNNH2N

CAS RN 2250-55-7

IUPAC Name: 6-(trifluoromethyl)-1H-indazol-3-amine
Synonyms: 6-(trifluoromethyl)-1H-indazol-3-amine
Molecular Weight (g/mol): 201.15
Molecular Formula: C8H6F3N3
InChi Key: ZMPFPQFXUYYHSJ-UHFFFAOYSA-N
SMILES: NC1=NNC2=CC(=CC=C12)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results