CAS RN 22852-13-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ N-Acetylpuromycin
Downregulates SnoN and Ski protein expression; promotes TGF-β signaling