CAS RN 228559-41-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ VEGFR2 Kinase Inhibitor VI, Ki8751
The VEGFR2 Kinase Inhibitor VI, Ki8751, also referenced under CAS 228559-41-9, controls the biological activity of VEGFR2 Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
2
Tocris Bioscience™ Ki 8751
Potent, selective VEGFR-2 inhibitor