Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ki 8751
GREENER_CHOICESDP

Catalog No. 254250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-425-0 50 mg
25-421-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-425-0 Supplier Tocris Bioscience™ Supplier No. 2542/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective VEGFR-2 inhibitor

Ki 8751 is a potent, selective inhibitor of VEGFR-2 tyrosine kinase (IC50 = 0.9 nM). Displays some inhibitory activity towards c-Kit, PDGFRα and FGFR-2 (IC50 values range from 40 to 170 nM) but is highly selective over other receptor tyrosine kinases (IC50 > 10000 nM for FGFR-2, EGFR and HGFR). Inhibits VEGF-stimulated proliferation of human umbilical vein endothelial cells (HUVEC) and inhibits tumor growth in vivo; antiangiogenic.

Specifications

Chemical Name or Material N-(2,4-Difluorophenyl)-N'-[4-[(6,7-dimethoxy-4-quinolinyl)oxy]-2-fluorophenyl]urea
CAS 228559-41-9
Assay Percent Range >97%
Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C24H18F3N3O4
Formula Weight Observed MW: 469.41

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.