accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(S)NH(R)HO(R)OH3CO

CAS RN 22862-76-6

IUPAC Name: (2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate
Synonyms: flagecidin upjohn 204t3 antibiotic PA-106
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: YKJYKKNCCRKFSL-BFHYXJOUSA-N
SMILES: COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1
Physical Form 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Type 
  • (4)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)