Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Anisomycin, 10 mg/ml in DMSO, sterile-filtered

Catalog No. AAJ67498XF
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67498XF 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67498XF Supplier Thermo Scientific Chemicals Supplier No. J67498XF
Add to Cart

CAS: 22862-76-6 | C14H19NO4 | 265.31 g/mol

It is applied as an antiparasitic agent and antineoplastic agent. It acts as a protein synthesis inhibitor (blocks translation), potent activator of stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase. It also scts as a potent signaling agonist to selectively elicit homologous desensitization of immediate early gene induction (c-fos, fosB, c-jun, junB and junD). Activates mitogen-activated protein (MAP) kinases (JNK/SAPK and p38/RK).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an antiparasitic agent and antineoplastic agent. It acts as a protein synthesis inhibitor (blocks translation), potent activator of stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase. It also scts as a p
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22862-76-6
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00077650
InChI Key YKJYKKNCCRKFSL-BFHYXJOUSA-N
Synonym Flagecidin
PubChem CID 253602
ChEBI CHEBI:338412
IUPAC Name (2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate
SMILES COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1

Specifications

Type Anisomycin
Physical Form Liquid
Quantity 1 mL
Solubility Information Fully miscible in water. Soluble to 50 mM in ethanol and to 100 mM in DMSO. Soluble in methanol (20mg/ml clear,faint yellow solution).Slightly soluble in alcohols,esters,ketones,chloroform,benzene,toluene,hexane,and ethyl acetate (10mg/ml clear,colorless solution),DMF (20mg/ml),and PBS pH 7.2 (0.5mg/ml).
Chemical Name or Material Anisomycin, 10mg/mL in DMSO, sterile-filtered
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 245-269-7
RTECSNumber BZ9800000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.