accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NHOHNOHNHOSONHONHOOHNH2NOONOHOHNH3COHOHONH

CAS RN 23109-05-9

IUPAC Name: 2-[34-(butan-2-yl)-13-(3,4-dihydroxybutan-2-yl)-8,22-dihydroxy-2,5,11,14,27,30,33,36,39-nonaoxo-27λ⁴-thia-3,6,12,15,25,29,32,35,38-nonaazapentacyclo[14.12.11.0⁶,¹⁰.0¹⁸,²⁶.0¹⁹,²⁴]nonatriaconta-18(26),19,21,23-tetraen-4-yl]acetamide
Synonyms: α-amanitin
Molecular Weight (g/mol): 918.98
Molecular Formula: C39H54N10O14S
InChi Key: CIORWBWIBBPXCG-UHFFFAOYNA-N
SMILES: CCC(C)C1NC(=O)CNC(=O)C2CC3=C(NC4=CC(O)=CC=C34)S(=O)CC(NC(=O)CNC1=O)C(=O)NC(CC(N)=O)C(=O)N1CC(O)CC1C(=O)NC(C(C)C(O)CO)C(=O)N2
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)