accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23138-64-9

CH3ONCO

CAS RN 23138-64-9

IUPAC Name: 1-(3-isocyanatophenyl)ethan-1-one
Synonyms: 1-(3-isocyanatophenyl)ethanone
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: JASTZOAHAWBRLM-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(=C1)N=C=O
Boiling Point 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results