CAS RN 2319922-08-0
CAS RN 2319922-08-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ ML 218 hydrochloride
CAS: 2319922-08-0 Molecular Formula: C19H27Cl3N2O