accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOHNH3COHOHNOH(R)(R)OHHOOOHO

CAS RN 23210-58-4

IUPAC Name: (2R,3R)-2,3-dihydroxybutanedioic acid; bis(4-[2-(4-benzylpiperidin-1-yl)-1-hydroxypropyl]phenol)
Synonyms: unii-89CTB4XUF7|ifenprodil tartrate|ifenprodil L-(+)-tartrate
Molecular Weight (g/mol): 800.99
Molecular Formula: C46H60N2O10
InChi Key: —
SMILES: O[C@H]([C@@H](O)C(O)=O)C(O)=O.CC(C(O)C1=CC=C(O)C=C1)N1CCC(CC2=CC=CC=C2)CC1.CC(C(O)C1=CC=C(O)C=C1)N1CCC(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results