CAS RN 2322286-81-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ DMHBO+
Fluorescent upon binding to Chili aptamer; for imaging RNA in cells