CAS RN 2341833-14-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ uPSEM 817 tartrate
Highly potent and selective PSAM4-GlyR and PSAM4-5-HT3 chimeric ion channel agonist; brain-penetrant