accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2362-63-2

CH3NH2S

CAS RN 2362-63-2

IUPAC Name: 3-methylbenzene-1-carbothioamide
Synonyms: 3-methylbenzenecarbothioamide
Molecular Weight (g/mol): 151.23
Molecular Formula: C8H9NS
InChi Key: NUFFXGAGGYWFAV-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(N)=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results