CAS RN 2375740-98-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ CSN5i-3
Potent and selective CSN5 (COP9 signalosome) inhibitor