CAS RN 24003-58-5
IUPAC Name:
{[(2,6-dimethylphenyl)carbamoyl]methyl}triethylazanium bromide
Synonyms:
{[(2,6-dimethylphenyl)carbamoyl]methyl}triethylazanium bromide
Molecular Weight (g/mol):
343.31
Molecular Formula:
C16H27BrN2O
InChi Key:
DLHMKHREUTXMCH-UHFFFAOYSA-N
SMILES:
[Br-].CC[N+](CC)(CC)CC(=O)NC1=C(C)C=CC=C1C
1
–
1
of
1
results
1
Tocris Bioscience™ QX 314 bromide
CAS: 24003-58-5 Molecular Formula: C16H27BrN2O