accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONCS

CAS RN 24066-82-8

IUPAC Name: ethyl 2-isothiocyanatoacetate
Synonyms: ethyl 2-isothiocyanatoacetate
Molecular Weight (g/mol): 145.18
Molecular Formula: C5H7NO2S
InChi Key: IYPSSPPKMLXXRN-UHFFFAOYSA-N
SMILES: CCOC(=O)CN=C=S
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)