CAS RN 2408341-97-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ VH 101, acid
VHL ligand, carboxylic acid functionalized at the benzylic position