Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
VH 101, acid is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand with carboxylic acid handle at the benzylic position ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | -3-[[[(2 S,4 R)-1-[-2-(1-Fluorocyclopropan-1-ylcarbonylamino)-3,3-dimethylbutanoyl]-4-hydroxypyrrolidin-2-yl]carbonyl]amino]-3-[4-(4-methylthiazol-5-yl)phenyl]propanoic acid |
| CAS | 2408341-97-7 |
| Quantity | 50 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C28H35FN4O6S |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

