Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mycophenolic acid
GREENER_CHOICESDP

Catalog No. 1505100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-051-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-051-00 Supplier R&D Systems™ Supplier No. 1505/100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inosine monophosphatase dehydrogenase inhibitor

Immunosuppressive agent with antiviral and antitumor effects in vitro and in vivo. Potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. Reversibly inhibits proliferation of T and B lymphocytes and antibody formation.

Chemical Identifiers

CAS 24280-93-1
Molecular Formula C17H20O6

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 320.34
Purity >98%
Molecular Weight (g/mol) 320.341
Chemical Name or Material Mycophenolic acid
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.