CAS RN 243984-11-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ TLR4 Inhibitor, TAK-242
TAK-242; CAS 243984-11-4; is a cell-permeable compound that selectively binds to Cys747 of TLR4 and selectively disrupts its interaction with adaptor molecules TIRAP and TRAM.
2
Tocris Bioscience™ TAK 242
Toll-like receptor 4 (TLR4) signaling inhibitor