accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OBrSOOHNO(E/Z)

CAS RN 244101-02-8

IUPAC Name: N-(5-bromo-2-methoxybenzenesulfonyl)-3-{2-[(naphthalen-2-yl)methyl]phenyl}prop-2-enamide
Synonyms: N-(5-bromo-2-methoxybenzenesulfonyl)-3-[2-(naphthalen-2-ylmethyl)phenyl]prop-2-enamide
Molecular Weight (g/mol): 536.44
Molecular Formula: C27H22BrNO4S
InChi Key: ODTKFNUPVBULRJ-UHFFFAOYSA-N
SMILES: COC1=C(C=C(Br)C=C1)S(=O)(=O)NC(=O)C=CC1=CC=CC=C1CC1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)