CAS RN 2446474-10-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ A 410099.1 amide-PEG4-amine
IAP ligand with PEG linker and terminal amine for onward chemistry