Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ A 410099.1 amide-PEG4-amine
GREENER_CHOICESDP

Catalog No. 722025 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-202-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 72-202-5 Supplier R&D Systems™ Supplier No. 7220/25

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. IAP ligand with PEG linker and terminal amine for onward chemistry

A 410099.1 amide-PEG4-amine is a functionalized IAP ligand for PROTAC™ research and development; incorporates an IAP ligand plus an amide-PEG4 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

Specifications

Chemical Name or Material tert-Butyl (-1-((-2-((2 S,4 S)-4-(14-amino-3,6,9,12-tetraoxatetradecanamido)-2-(((R)-1,2,3,4-tetrahydronaphthalen-1-yl)carbamoyl)pyrrolidin-1-yl)-1-cyclohexyl-2-oxoethyl)amino)-1-oxopropan-2-yl)(methyl)carbamate
CAS 2446474-10-6
Quantity 25 mg
Product Line Tocris
Target Degrader Building Blocks
Molecular Formula C42H68N6O10
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.