CAS RN 2459-05-4
CAS RN 2459-05-4
IUPAC Name:
(2E)-4-ethoxy-4-oxobut-2-enoic acid
Synonyms:
(2E)-4-ethoxy-4-oxobut-2-enoic acid
Molecular Weight (g/mol):
144.13
Molecular Formula:
C6H8O4
InChi Key:
XLYMOEINVGRTEX-ONEGZZNKSA-N
SMILES:
CCOC(=O)\C=C\C(O)=O
1
–
1
of
1
results
1
Monoethyl Fumarate 97.0+%, TCI America™
CAS: 2459-05-4 Molecular Formula: C6H8O4 Molecular Weight (g/mol): 144.13 MDL Number: MFCD00002699 InChI Key: XLYMOEINVGRTEX-ONEGZZNKSA-N Synonym: monoethyl fumarate,e-4-ethoxy-4-oxobut-2-enoic acid,ethyl hydrogen fumarate,fumaric acid monoethyl ester,e-4-ethoxy-4-oxo-2-butenoic acid,unii-y3w849ng2n,2e-4-ethoxy-4-oxobut-2-enoic acid,fumaric acid, monoethyl ester,mono-ethyl fumarate,ethyl hydrogen maleate PubChem CID: 5358902 IUPAC Name: (E)-4-ethoxy-4-oxobut-2-enoic acid SMILES: CCOC(=O)C=CC(=O)O