accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2521-07-5

CH3NN

CAS RN 2521-07-5

IUPAC Name: 9-methyl-9H-pyrido[3,4-b]indole
Synonyms: 9-methylpyrido[3,4-b]indole
Molecular Weight (g/mol): 182.23
Molecular Formula: C12H10N2
InChi Key: MABOIYXDALNSES-UHFFFAOYSA-N
SMILES: CN1C2=C(C=CC=C2)C2=C1C=NC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results