accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFNN

CAS RN 25371-96-4

IUPAC Name: 1-[2-(trifluoromethyl)phenyl]-1H-imidazole
Synonyms: 1-[2-(trifluoromethyl)phenyl]imidazole
Molecular Weight (g/mol): 212.18
Molecular Formula: C10H7F3N2
InChi Key: WZBWBNCQUTXYEL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1N1C=CN=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)