accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OHOOHNO

CAS RN 2552-55-8

IUPAC Name: 2-amino-2-(3-oxo-2,3-dihydro-1,2-oxazol-5-yl)acetic acid
Synonyms: ibotenic acid
Molecular Weight (g/mol): 158.11
Molecular Formula: C5H6N2O4
InChi Key: IRJCBFDCFXCWGO-UHFFFAOYNA-N
SMILES: NC(C(O)=O)C1=CC(=O)NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)