CAS RN 256373-96-3
IUPAC Name:
sodium [(1,2,3,5,6,7-hexahydro-s-indacen-4-yl)carbamoyl]({[4-(2-hydroxypropan-2-yl)furan-2-yl]sulfonyl})azanide
Synonyms:
sodium [(1,2,3,5,6,7-hexahydro-s-indacen-4-yl)carbamoyl][4-(2-hydroxypropan-2-yl)furan-2-ylsulfonyl]azanide
Molecular Weight (g/mol):
426.46
Molecular Formula:
C20H23N2NaO5S
InChi Key:
LFQQNXFKPNZRFT-UHFFFAOYSA-M
SMILES:
[Na+].CC(C)(O)C1=COC(=C1)S(=O)(=O)[N-]C(=O)NC1=C2CCCC2=CC2=C1CCC2