accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNNOFFFFFF

CAS RN 256414-75-2

IUPAC Name: 5-[4-phenyl-5-(trifluoromethyl)thiophen-2-yl]-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole
Synonyms: 5-[4-phenyl-5-(trifluoromethyl)thiophen-2-yl]-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole
Molecular Weight (g/mol): 440.36
Molecular Formula: C20H10F6N2OS
InChi Key: OYMNPJXKQVTQTR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(C=C(S1)C1=NC(=NO1)C1=CC(=CC=C1)C(F)(F)F)C1=CC=CC=C1
Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results