CAS RN 2567869-47-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
R&D Systems™ MRS 2768 tetrasodium salt
CAS: 2567869-47-8 Molecular Formula: C15H16N2Na4O18P4