accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2568-30-1

ClOO

CAS RN 2568-30-1

IUPAC Name: 2-(chloromethyl)-1,3-dioxolane
Synonyms: 2-(chloromethyl)-1,3-dioxolane
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: IKZOMJGRWIOEDP-UHFFFAOYSA-N
SMILES: ClCC1OCCO1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)