accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNHOOHCH3OOHOOH(E/Z)

CAS RN 257603-40-0

IUPAC Name: 2-cyclopentyl-2-hydroxy-N-[1-(4-methylpent-3-en-1-yl)piperidin-4-yl]-2-phenylacetamide; but-2-enedioic acid
Synonyms: 2-cyclopentyl-2-hydroxy-N-[1-(4-methylpent-3-en-1-yl)piperidin-4-yl]-2-phenylacetamide; butenedioic acid
Molecular Weight (g/mol): 500.64
Molecular Formula: C28H40N2O6
InChi Key: HUCQUCITPHOUAC-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.CC(C)=CCCN1CCC(CC1)NC(=O)C(O)(C1CCCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results