accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2610-05-1

IUPAC Name: tetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2Z)-8-amino-1-oxo-5,7-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxo-5,6-dihydronaphthalene-1,3-disulfonate
Synonyms: tetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2Z)-8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxonaphthalene-1,3-disulfonate direct blue 1
Molecular Weight (g/mol): 992.79
Molecular Formula: C34H24N6Na4O16S4
InChi Key: UPKAWFACSJWKND-ZXFFUEEESA-J
SMILES: [Na+].[Na+].[Na+].[Na+].COC1=CC(=CC=C1N\N=C1\C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(OC)=C1
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results