CAS RN 265129-71-3
IUPAC Name:
2-[(4-{2-[(4-cyclohexylbutyl)(cyclohexylcarbamoyl)amino]ethyl}phenyl)sulfanyl]-2-methylpropanoic acid
Synonyms:
2-[(4-{2-[(4-cyclohexylbutyl)(cyclohexylcarbamoyl)amino]ethyl}phenyl)sulfanyl]-2-methylpropanoic acid
Molecular Weight (g/mol):
502.76
Molecular Formula:
C29H46N2O3S
InChi Key:
PKNYXWMTHFMHKD-UHFFFAOYSA-N
SMILES:
CC(C)(SC1=CC=C(CCN(CCCCC2CCCCC2)C(=O)NC2CCCCC2)C=C1)C(O)=O
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ GW7647
A cell-permeable urea-substituted thioisobutyric acid compound that acts as a potent and selective PPARα.
2
Tocris Bioscience™ GW 7647
CAS: 265129-71-3 Molecular Formula: C29H46N2O3S