Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GW 7647
GREENER_CHOICESDP

Catalog No. 167750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-775-0 50 mg
16-771-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-775-0 Supplier Tocris Bioscience™ Supplier No. 1677/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly selective, potent PPARα agonist. Orally active

Potent and highly selective PPARα agonist (EC50 values are 6, 1100 and 6200 nM for human PPARα, PPARγ and PPARδ receptors respectively). Modulates oleate metabolism and mitochondrial enzyme gene expression in mature myotubules in vitro. Has lipid-lowering effects following oral administration in vivo. Reduces NO production in macrophages; exhibits anti-inflammatory properties.

Chemical Identifiers

CAS 265129-71-3
Molecular Formula C29H46N2O3S

Specifications

Quantity 50 mg
Formula Weight 502.75
Purity >99%
Molecular Weight (g/mol) 502.76
Chemical Name or Material GW 7647
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.