accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 265312-55-8

IUPAC Name: 2-methyl-5-[(4-methylphenyl)amino]-4,7-dihydro-1,3-benzothiazole-4,7-dione
Synonyms: 2-methyl-5-[(4-methylphenyl)amino]-1,3-benzothiazole-4,7-dione
Molecular Weight (g/mol): 284.33
Molecular Formula: C15H12N2O2S
InChi Key: HFPLHASLIOXVGS-UHFFFAOYSA-N
SMILES: CC1=NC2=C(S1)C(=O)C=C(NC1=CC=C(C)C=C1)C2=O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results