accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26939-18-4

H3CH3CCH3NCH3O

CAS RN 26939-18-4

IUPAC Name: 2,4,4,6-tetramethyl-5,6-dihydro-4H-1,3-oxazine
Synonyms: 2,4,4,6-tetramethyl-5,6-dihydro-1,3-oxazine
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H15NO
InChi Key: LZSGJOXDYPFFQB-UHFFFAOYNA-N
SMILES: CC1CC(C)(C)N=C(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results