accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOH3CCH3CH3

CAS RN 27043-27-2

IUPAC Name: 1-(2,3-dimethylbenzenesulfonyl)-2,3-dimethylbenzene
Synonyms: 1-(2,3-dimethylbenzenesulfonyl)-2,3-dimethylbenzene
Molecular Weight (g/mol): 274.38
Molecular Formula: C16H18O2S
InChi Key: IOGPZWCEELGKFR-UHFFFAOYSA-N
SMILES: CC1=C(C)C(=CC=C1)S(=O)(=O)C1=CC=CC(C)=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results