CAS RN 273404-37-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ VX 765
High affinity and selective caspase-4 and caspase 1/ICE inhibitor; orally bioavailable
2
MilliporeSigma™ Calbiochem™ Caspase-1/4 Inhibitor, VX-765
A cell-permeable prodrug that upon conversion to its active form inhibits caspase-1 and 4 activity (Ki = 800 pM and 600 pM, respectively).