CAS RN 2734853-80-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ ZYS 1
ADAR1 (adenosine deaminase acting on RNA 1) deaminase inhibitor