CAS RN 2734909-87-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ c-Di-AMP sodium salt
Endogenous STING and DDX41 agonist; activates STING-dependent signaling