Learn More
Description
This product is sold in units of 702μg, equivalent to 1μmol.
Specifications
Specifications
| Chemical Name or Material | 3′,5′-Cyclic diadenylic acid sodium salt |
| CAS | 2734909-87-4 |
| Quantity | 702 μg |
| Product Line | Tocris |
| Target | STING Antagonists |
| Molecular Formula | C20H22N10Na2O12P2 |
| Formula Weight | 702.38 |
| Purity | >98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

