CAS RN 2740620-18-0
CAS RN 2740620-18-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ BDY FL Thalidomide
High affinity fluorescent probe for TR-FRET cereblon binding assay