accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2747-05-9

H3COOOOCH3

CAS RN 2747-05-9

IUPAC Name: 4-methyl-2-oxo-2H-chromen-7-yl acetate
Synonyms: 4-methylumbelliferyl acetate β-methylumbelliferyl acetate
Molecular Weight (g/mol): 218.21
Molecular Formula: C12H10O4
InChi Key: HXVZGASCDAGAPS-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C2C(C)=CC(=O)OC2=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)