accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2768-02-7

CH3OSiOH3COH3CH2C

CAS RN 2768-02-7

IUPAC Name: ethenyltrimethoxysilane
Synonyms: silane, ethenyltrimethoxy-
Molecular Weight (g/mol): 148.23
Molecular Formula: C5H12O3Si
InChi Key: NKSJNEHGWDZZQF-UHFFFAOYSA-N
SMILES: CO[Si](OC)(OC)C=C
Molecular Weight (g/mol) 
  • (4)
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (1)
  • (3)
  • (4)
  • (4)
  • (1)
  • (3)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (5)