accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 286-62-4

IUPAC Name: 9-oxabicyclo[6.1.0]nonane
Synonyms: 9-oxabicyclo[6.1.0]nonane
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: MELPJGOMEMRMPL-UHFFFAOYNA-N
SMILES: C1CCCC2OC2CC1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)