CAS RN 28643-80-3
IUPAC Name:
sodium 2-[(3S,6R)-6-{[(5R,7R,9R,10R)-2-[(3'S,5'R)-5'-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-2,3'-dimethyl-[2,2'-bioxolan]-5-yl]-9-methoxy-2,4,10-trimethyl-1,6-dioxaspiro[4.5]decan-7-yl]methyl}-3-methyloxan-2-yl]propanoate
Synonyms:
sodium 2-[(3S,6R)-6-{[(5R,7R,9R,10R)-2-[(3'S,5'R)-5'-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-2,3'-dimethyl-[2,2'-bioxolan]-5-yl]-9-methoxy-2,4,10-trimethyl-1,6-dioxaspiro[4.5]decan-7-yl]methyl}-3-methyloxan-2-yl]propanoate
Molecular Weight (g/mol):
746.96
Molecular Formula:
C40H67NaO11
InChi Key:
MOYOTUKECQMGHE-KKCUGXASSA-M
SMILES:
[Na+].CO[C@@H]1C[C@@H](C[C@H]2CC[C@H](C)C(O2)C(C)C([O-])=O)O[C@]2(OC(C)(CC2C)C2CCC(C)(O2)C2O[C@H](C[C@@H]2C)[C@H]2O[C@@](O)(CO)[C@H](C)C[C@@H]2C)[C@@H]1C
1
–
2
of
2
results
2
R&D Systems™ Nigericin sodium salt
CAS: 28643-80-3 Molecular Formula: C40H67NaO11