accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)OHHONHH2NOHO

CAS RN 28860-95-9

IUPAC Name: (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid
Synonyms: carbidopa carbidopa, (r)-isomer α-methyldopa hydrazine carbidopa (anhydrous) (S)-carbidopa lodosin lodosyn carbidopa monohydrate hadrazino-α-methyldopa (s)-carbidopa
Molecular Weight (g/mol): 226.23
Molecular Formula: C10H14N2O4
InChi Key: TZFNLOMSOLWIDK-JTQLQIEISA-N
SMILES: C[C@@](CC1=CC=C(O)C(O)=C1)(NN)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results