CAS RN 289905-88-0
IUPAC Name:
1-[(2-chlorophenyl)diphenylmethyl]-1H-pyrazole
Synonyms:
tram-34
Molecular Weight (g/mol):
344.84
Molecular Formula:
C22H17ClN2
InChi Key:
KBFUQFVFYYBHBT-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC=C1C(N1C=CC=N1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals TRAM 34
Thermo Scientific Chemicals TRAM 34
CAS: 289905-88-0 Molecular Formula: C22H17ClN2 Synonym: 1-[(2-Chlorophenyl)diphenylmethyl]-1H-pyrazole
2
R&D Systems™ TRAM 34
CAS: 289905-88-0 Molecular Formula: C22H17ClN2