CAS RN 2901113-25-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ JP-2-196 alkyne
RNF126 chemical handle; alkyne functionalized version of JP-2-196 (Cat. No. 8046).